Assay Details
Target Gene Details
Entrez Gene ID: | 196500 |
Gene Name: | PILR alpha associated neural protein |
Gene Aliases: |
C12orf53, LEDA1, PANP, leda-1 |
Location: |
Chr.12:6689726-6700843 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIANP | NM_001244014.1 | NP_001230943.1 | ||
| NM_001244015.1 | NP_001230944.1 | |||
| NM_153685.3 | NP_710152.1 | |||
| XM_011520926.2 | XP_011519228.1 | |||
| AK056295.1 | ||||
| AK091596.1 | ||||
| AK091656.1 | ||||
| AK095854.1 | ||||
| AK289648.1 | ||||
| AY358534.1 | AAQ88898.1 | |||
| BC035736.1 | AAH35736.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3580394 | Chr.12:6696040 - 6779214 on Build GRCh38 | Gain |
|
| esv2761725 | Chr.12:6682182 - 6754772 on Build GRCh38 | Gain |
|
| nsv832322 | Chr.12:6575884 - 6765914 on Build GRCh38 | Loss |
|
| nsv832323 | Chr.12:6662233 - 6856559 on Build GRCh38 | Loss |
|

Genomic Map