Assay Details
Target Gene Details
Entrez Gene ID: | 51720 |
Gene Name: | ubiquitin interaction motif containing 1 |
Gene Aliases: |
RAP80, X2HRIP110 |
Location: |
Chr.5:176905005-177022640 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UIMC1 | NM_001199297.2 | 13 | 2145 | NP_001186226.1 |
| NM_001199298.1 | 12 | 1920 | NP_001186227.1 | |
| NM_001317961.1 | 12 | 1533 | NP_001304890.1 | |
| NM_016290.4 | 12 | 1934 | NP_057374.3 | |
| XM_005265930.2 | 12 | 1908 | XP_005265987.1 | |
| XM_005265932.2 | 10 | 1044 | XP_005265989.1 | |
| XM_005265933.2 | 10 | 1156 | XP_005265990.1 | |
| XM_005265934.2 | 12 | 1363 | XP_005265991.1 | |
| XM_005265935.2 | 12 | 1372 | XP_005265992.1 | |
| XM_005265936.2 | 11 | 1162 | XP_005265993.1 | |
| XM_006714871.2 | 12 | 1883 | XP_006714934.1 | |
| XM_006714874.2 | 12 | 1347 | XP_006714937.1 | |
| XM_011534568.2 | XP_011532870.1 | |||
| XM_017009574.1 | 11 | 1799 | XP_016865063.1 | |
| XM_017009575.1 | 13 | 1611 | XP_016865064.1 | |
| XM_017009576.1 | 10 | 1057 | XP_016865065.1 | |
| XM_017009577.1 | 10 | 1109 | XP_016865066.1 | |
| XM_017009578.1 | 11 | 1171 | XP_016865067.1 | |
| XM_017009579.1 | 11 | 1071 | XP_016865068.1 | |
| AF113538.1 | 8 | 724 | AAF14875.1 | |
| AF349313.1 | 12 | 1876 | AAK61871.1 | |
| AK023044.1 | 13 | 1922 | ||
| AL137375.1 | 3 | 252 | CAB70716.1 | |
| BC006078.1 | 12 | 1841 | AAH06078.1 | |
| BC032561.1 | 12 | 1324 | AAH32561.1 | |
| BX537376.1 | 12 | 1934 | CAD97618.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1016283 | Chr.5:176904803 - 176968410 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map