Assay Details
Target Gene Details
Entrez Gene ID: | 152330 |
Gene Name: | contactin 4 |
Gene Aliases: |
AXCAM, BIG-2 |
Location: |
Chr.3:2098771-3059080 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 31 - Intron 31 |
Target Gene Details
Entrez Gene ID: | 100873975 |
Gene Name: | CNTN4 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:3039661-3061145 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNTN4-AS1 | NR_046554.1 | |||
| BM920658.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv589371 | Chr.3:2170634 - 3594980 on Build GRCh38 | Loss |
|
| esv2751994 | Chr.3:2200757 - 3178886 on Build GRCh38 | Gain |
|
| esv2422274 | Chr.3:2100396 - 3314907 on Build GRCh38 | Duplication |
|
| nsv1011872 | Chr.3:2782874 - 3057776 on Build GRCh38 | Gain |
|
| nsv521796 | Chr.3:3038825 - 3151219 on Build GRCh38 | Loss |
|
| dgv4636n100 | Chr.3:2825734 - 3271845 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map