Assay Details
Target Gene Details
Entrez Gene ID: | 58158 |
Gene Name: | neuronal differentiation 4 |
Gene Aliases: |
ATH-3, ATH3, Atoh3, MATH-3, MATH3, bHLHa4 |
Location: |
Chr.12:55019945-55030017 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEUROD4 | NM_021191.2 | 2 | 453 | NP_067014.2 |
| AK314136.1 | 2 | 368 | ||
| DA417566.1 | 2 | 368 |
More Information
Set Membership: |
|

Genomic Map