Assay Details
Target Gene Details
Entrez Gene ID: | 390442 |
Gene Name: | olfactory receptor family 11 subfamily H member 4 |
Gene Aliases: |
OR14-36 |
Location: |
Chr.14:20242792-20243766 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR11H4 | NM_001004479.1 | 1 | 230 | NP_001004479.1 |
| BC137054.1 | 1 | 283 | ||
| BC137055.1 | 1 | 283 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1797n100 | Chr.14:20218848 - 20275110 on Build GRCh38 | Loss |
|
| esv3633681 | Chr.14:20213255 - 20257091 on Build GRCh38 | Loss |
|
| nsv832739 | Chr.14:20135967 - 20266063 on Build GRCh38 | Gain |
|
| nsv563845 | Chr.14:20216118 - 20244163 on Build GRCh38 | Gain |
|
| esv3633679 | Chr.14:20210486 - 20289540 on Build GRCh38 | Loss |
|
| esv3633682 | Chr.14:20239929 - 20276665 on Build GRCh38 | Loss |
|
| esv2751268 | Chr.14:19734966 - 20248682 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map