Assay Details
Target Gene Details
Entrez Gene ID: | 1195 |
Gene Name: | CDC like kinase 1 |
Gene Aliases: |
CLK, CLK/STY, STY |
Location: |
Chr.2:200853009-200864744 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLK1 | NM_001162407.1 | 2 | 538 | NP_001155879.1 |
| NM_004071.3 | 2 | 258 | NP_004062.2 | |
| NR_027855.1 | 2 | 258 | ||
| NR_027856.1 | 2 | 258 | ||
| AK289365.1 | 2 | 210 | ||
| AK294295.1 | 2 | 538 | ||
| AK307301.1 | 2 | 172 | ||
| AK316127.1 | 2 | 229 | ||
| BC028149.1 | 2 | 154 | ||
| BC031549.1 | 2 | 154 | AAH31549.1 | |
| BE241788.1 | 2 | 216 | ||
| DA225472.1 | 2 | 258 | ||
| L29219.1 | 2 | 232 | AAA61480.1 | |
| L29222.1 | 2 | 232 | AAB59459.1 | |
| M59287.1 | 2 | 232 |
More Information
Set Membership: |
|

Genomic Map