Assay Details
Target Gene Details
Entrez Gene ID: | 339761 |
Gene Name: | cytochrome P450 family 27 subfamily C member 1 |
Gene Aliases: |
- |
Location: |
Chr.2:127183832-127220096 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP27C1 | NM_001001665.3 | NP_001001665.3 | ||
| XM_017003960.1 | XP_016859449.1 | |||
| XM_017003961.1 | XP_016859450.1 | |||
| AK131190.1 | BAD18388.1 | |||
| BC039307.1 | AAH39307.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4066n100 | Chr.2:127072541 - 127274970 on Build GRCh38 | Gain |
|
| esv3592352 | Chr.2:127170604 - 127210940 on Build GRCh38 | Gain |
|
| esv2759090 | Chr.2:127073367 - 127248938 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map