Assay Details
Target Gene Details
Entrez Gene ID: | 6038 |
Gene Name: | ribonuclease A family member 4 |
Gene Aliases: |
RAB1, RNS4 |
Location: |
Chr.14:20684213-20700602 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNASE4 | NM_001282192.1 | NP_001269121.1 | ||
| NM_001282193.1 | NP_001269122.1 | |||
| NM_002937.4 | NP_002928.1 | |||
| NM_194431.2 | NP_919412.1 | |||
| AK092422.1 | ||||
| BC015520.1 | AAH15520.1 | |||
| BG616902.1 | ||||
| BU594423.1 | ||||
| BX247985.1 | CAD62319.1 | |||
| BX248010.1 | CAD62338.1 | |||
| D37931.1 | BAA07150.1 | |||
| DA925725.1 | ||||
| DR003809.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3633688 | Chr.14:20684511 - 20707081 on Build GRCh38 | Loss |
|
| esv23438 | Chr.14:20332606 - 20789719 on Build GRCh38 | Gain+Loss |
|
| esv3633687 | Chr.14:20683687 - 20720960 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map