Assay Details
Target Gene Details
Entrez Gene ID: | 917 |
Gene Name: | CD3g molecule |
Gene Aliases: |
CD3-GAMMA, IMD17, T3G |
Location: |
Chr.11:118344307-118353782 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD3G | NM_000073.2 | NP_000064.1 | ||
| XM_005271724.3 | XP_005271781.1 | |||
| XM_006718941.2 | XP_006719004.1 | |||
| AK223314.1 | BAD97034.1 | |||
| AK299997.1 | ||||
| AK310578.1 | ||||
| AK310622.1 | ||||
| AK313966.1 | ||||
| AK315895.1 | ||||
| BC113830.1 | AAI13831.1 | |||
| DC378393.1 | ||||
| X04145.1 | CAA27764.1 | |||
| X60492.1 |
More Information
Set Membership: |
|

Genomic Map