Assay Details
Target Gene Details
Entrez Gene ID: | 117194 |
Gene Name: | MAS related GPR family member X2 |
Gene Aliases: |
MGRG3, MRGX2 |
Location: |
Chr.11:19054455-19060681 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRGPRX2 | NM_054030.3 | NP_473371.1 | ||
| BC063450.1 | AAH63450.1 |
More Information
Set Membership: |
|

Genomic Map