Assay Details
Target Gene Details
Entrez Gene ID: | 3752 |
Gene Name: | potassium voltage-gated channel subfamily D member 3 |
Gene Aliases: |
BRGDA9, KCND3L, KCND3S, KSHIVB, KV4.3, SCA19, SCA22 |
Location: |
Chr.1:111770662-111989577 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCND3 | NM_004980.4 | 8 | 2444 | NP_004971.2 |
| NM_172198.2 | 7 | 2387 | NP_751948.1 | |
| XM_005270851.4 | 8 | 2109 | XP_005270908.1 | |
| XM_006710629.3 | 8 | 2419 | XP_006710692.1 | |
| XM_006710630.3 | 7 | 2052 | XP_006710693.1 | |
| XM_017001244.1 | 8 | 2434 | XP_016856733.1 | |
| AF048712.1 | AAC05121.1 | |||
| AF048713.1 | AAC05122.1 | |||
| AF120491.1 | 8 | 2452 | AAD38898.1 | |
| AF187963.1 | AAF01044.1 | |||
| AF187964.1 | AAF01045.1 | |||
| AF205856.1 | AAF20924.1 | |||
| AF205857.1 | AAF20925.1 | |||
| BC113475.1 | 6 | 1929 | ||
| BC113477.1 | 6 | 1929 | ||
| BC143390.1 | 6 | 1929 |
More Information
Set Membership: |
|

Genomic Map