Assay Details
Target Gene Details
Entrez Gene ID: | 7903 |
Gene Name: | ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 4 |
Gene Aliases: |
PST, PST1, SIAT8D, ST8SIA-IV |
Location: |
Chr.5:100806935-100903285 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ST8SIA4 | NM_005668.5 | 5 | 4447 | NP_005659.1 |
| XM_011543630.1 | 4 | 4153 | XP_011541932.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3890678 | Chr.5:100324445 - 101099725 on Build GRCh38 | Gain |
|
| nsv1023765 | Chr.5:100579307 - 101398051 on Build GRCh38 | Loss |
|
| nsv1021183 | Chr.5:100290153 - 100885722 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map