Assay Details
Target Gene Details
Entrez Gene ID: | 55732 |
Gene Name: | chromosome 1 open reading frame 112 |
Gene Aliases: |
- |
Location: |
Chr.1:169794730-169854080 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1orf112 | NM_001320051.1 | NP_001306980.1 | ||
| AK127098.1 |
Target Gene Details
Entrez Gene ID: | 92342 |
Gene Name: | methyltransferase like 18 |
Gene Aliases: |
AsTP2, C1orf156, HPM1 |
Location: |
Chr.1:169792524-169794976 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| METTL18 | NM_001320199.1 | NP_001307128.1 | ||
| NM_033418.3 | 1 | 85 | NP_219486.1 | |
| AK313909.1 | ||||
| AL035369.1 | CAA23019.1 | |||
| AL556604.3 | 1 | 75 | ||
| BG614785.1 | ||||
| HY159746.1 | 1 | 85 |
More Information
Set Membership: |
|

Genomic Map