Assay Details
Target Gene Details
Entrez Gene ID: | 29985 |
Gene Name: | solute carrier family 39 member 3 |
Gene Aliases: |
ZIP-3, ZIP3 |
Location: |
Chr.19:2732524-2740076 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC39A3 | NM_144564.4 | NP_653165.2 | ||
| NM_213568.1 | 2 | 2618 | NP_998733.1 | |
| AK002044.1 | ||||
| AK302600.1 | ||||
| BC005869.2 | AAH05869.2 | |||
| BC020571.2 | 2 | 2586 | AAH20571.1 | |
| BG719479.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3643437 | Chr.19:2733028 - 2744541 on Build GRCh38 | Gain |
|
| nsv509714 | Chr.19:2519629 - 2794403 on Build GRCh38 | Insertion |
|
| nsv828397 | Chr.19:2688987 - 2800506 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map