Assay Details
Target Gene Details
Entrez Gene ID: | 26287 |
Gene Name: | ankyrin repeat domain 2 |
Gene Aliases: |
ARPP |
Location: |
Chr.10:97572441-97583884 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD2 | NM_001129981.2 | NP_001123453.1 | ||
| NM_001291218.1 | NP_001278147.1 | |||
| NM_001291219.1 | NP_001278148.1 | |||
| NM_020349.3 | NP_065082.2 | |||
| AB058599.1 | BAB60958.1 | |||
| AJ249975.1 | CAB99416.1 | |||
| AJ304805.1 | CAC19412.1 | |||
| AJ583444.1 | CAE47432.1 | |||
| AK056990.1 | ||||
| AK314763.1 | ||||
| BC020817.1 | AAH20817.2 | |||
| BC107759.1 | AAI07760.1 | |||
| DA893829.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7509 | Chr.10:97557833 - 97586895 on Build GRCh38 | Insertion |
|
| nsv1036494 | Chr.10:97507882 - 97585450 on Build GRCh38 | Gain |
|
| nsv1047359 | Chr.10:97546744 - 97609256 on Build GRCh38 | Gain |
|
| nsv818782 | Chr.10:97577815 - 97590371 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map