Assay Details
Target Gene Details
Entrez Gene ID: | 339834 |
Gene Name: | coiled-coil domain containing 36 |
Gene Aliases: |
CT74 |
Location: |
Chr.3:49198428-49258104 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC36 | NM_001135197.1 | NP_001128669.1 | ||
| NM_178173.3 | NP_835467.2 | |||
| XM_006713124.3 | XP_006713187.1 | |||
| XM_011533670.2 | XP_011531972.1 | |||
| XM_011533671.2 | XP_011531973.1 | |||
| XM_011533672.2 | XP_011531974.1 | |||
| XM_011533673.2 | XP_011531975.1 | |||
| AK058049.1 | BAB71640.1 | |||
| AK303033.1 | ||||
| BC036202.1 | AAH36202.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1008750 | Chr.3:49171382 - 49324968 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map