Assay Details
Target Gene Details
Entrez Gene ID: | 101929710 |
Gene Name: | uncharacterized LOC101929710 |
Gene Aliases: |
- |
Location: |
Chr.5:95962001-96631085 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929710 | NR_130776.1 | |||
| DA871062.1 |
Target Gene Details
Entrez Gene ID: | 5122 |
Gene Name: | proprotein convertase subtilisin/kexin type 1 |
Gene Aliases: |
BMIQ12, NEC1, PC1, PC3, SPC3 |
Location: |
Chr.5:96390336-96433281 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCSK1 | NM_000439.4 | 14 | 2449 | NP_000430.3 |
| NM_001177875.1 | 14 | 2107 | NP_001171346.1 | |
| AB208874.1 | 14 | 2125 | BAD92111.1 | |
| AK303888.1 | 14 | 2107 | ||
| AK308575.1 | 8 | 1355 | ||
| BC130295.1 | 14 | 2331 | ||
| BC136486.1 | 14 | 2331 | ||
| M90753.1 | 14 | 2419 | AAA59918.1 | |
| X64810.1 | 14 | 2399 | CAA46031.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1021263 | Chr.5:96060753 - 97448132 on Build GRCh38 | Loss |
|
| esv3570258 | Chr.5:96392303 - 96394937 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map