Assay Details
Target Gene Details
Entrez Gene ID: | 79947 |
Gene Name: | dehydrodolichyl diphosphate synthase subunit |
Gene Aliases: |
CIT, CPT, DS, HDS, RP59 |
Location: |
Chr.1:26432282-26471306 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
More Information
Set Membership: |
|

Genomic Map