Assay Details
Target Gene Details
Entrez Gene ID: | 30836 |
Gene Name: | deoxynucleotidyltransferase terminal interacting protein 2 |
Gene Aliases: |
ERBP, FCF2, HSU15552, LPTS-RP2, TdIF2 |
Location: |
Chr.1:93869458-93879206 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNTTIP2 | NM_014597.4 | 2 | 1242 | NP_055412.2 |
| AB046574.1 | 2 | 1222 | BAD08331.1 | |
| AK123702.1 | 2 | 2483 | ||
| AK222722.1 | 2 | 1215 | BAD96442.1 | |
| AK292379.1 | 2 | 1215 | ||
| AK302909.1 | 2 | 1198 | ||
| AK308808.1 | 2 | 842 | ||
| AY336729.1 | 2 | 1223 | AAR02407.1 | |
| AY394925.1 | 2 | 1208 | AAQ95169.1 | |
| BC047688.1 | 2 | 1204 | AAH47688.1 | |
| BC130622.1 | 2 | 1220 | ||
| U15552.1 | 2 | 1208 | AAA50601.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv998151 | Chr.1:93876112 - 93877555 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map