Assay Details
Target Gene Details
Entrez Gene ID: | 11041 |
Gene Name: | beta-1,4-glucuronyltransferase 1 |
Gene Aliases: |
B3GN-T1, B3GNT1, B3GNT6, BETA3GNTI, MDDGA13, iGAT, iGNT |
Location: |
Chr.11:66345372-66347690 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B4GAT1 | NM_006876.2 | 2 | 1815 | NP_006867.1 |
| AF029893.1 | 2 | 1749 | AAC39538.1 | |
| AK075354.1 | 2 | 1699 | ||
| AK098389.1 | 2 | 1754 | ||
| AK098779.1 | 2 | 1699 | ||
| AK225182.1 | 2 | 1702 | ||
| BC021965.2 | 2 | 1739 | AAH21965.1 |
More Information
Set Membership: |
|

Genomic Map