Assay Details
Target Gene Details
Entrez Gene ID: | 391112 |
Gene Name: | olfactory receptor family 6 subfamily Y member 1 |
Gene Aliases: |
OR1-11, OR6Y2 |
Location: |
Chr.1:158547128-158548105 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR6Y1 | NM_001005189.1 | 1 | 212 | NP_001005189.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv515774 | Chr.1:158547563 - 158551602 on Build GRCh38 | Loss |
|
| dgv15n17 | Chr.1:158541655 - 158554646 on Build GRCh38 | Loss |
|
| esv3578210 | Chr.1:158547193 - 158550492 on Build GRCh38 | Loss |
|
| esv2667471 | Chr.1:158546972 - 158551953 on Build GRCh38 | Deletion |
|
| dgv444n100 | Chr.1:158547194 - 158575633 on Build GRCh38 | Gain |
|
| dgv443n100 | Chr.1:158545091 - 158556782 on Build GRCh38 | Loss |
|
| esv2764188 | Chr.1:158543258 - 158557734 on Build GRCh38 | Loss |
|
| esv2422163 | Chr.1:158547046 - 158551602 on Build GRCh38 | Deletion |
|
| nsv442095 | Chr.1:158547206 - 158550627 on Build GRCh38 | Loss |
|
| nsv470743 | Chr.1:158514495 - 158551602 on Build GRCh38 | Loss |
|
| esv3587675 | Chr.1:158547359 - 158551889 on Build GRCh38 | Loss |
|
| nsv547983 | Chr.1:158525674 - 158553881 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map