Assay Details
Target Gene Details
Entrez Gene ID: | 3034 |
Gene Name: | histidine ammonia-lyase |
Gene Aliases: |
HIS, HSTD |
Location: |
Chr.12:95972662-95996365 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HAL | NM_001258333.1 | 20 | 2940 | NP_001245262.1 |
| NM_001258334.1 | 20 | 2937 | NP_001245263.1 | |
| NM_002108.3 | 21 | 3007 | NP_002099.1 | |
| XM_011538249.2 | 12 | 1840 | XP_011536551.1 | |
| XM_017019246.1 | 12 | 2328 | XP_016874735.1 | |
| D16626.1 | 21 | 2881 | BAA04047.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs113875416] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map