Assay Details
Target Gene Details
Entrez Gene ID: | 140902 |
Gene Name: | R3H domain containing like |
Gene Aliases: |
dJ881L22.3 |
Location: |
Chr.20:44337158-44350792 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| R3HDML | NM_178491.3 | NP_848586.1 | ||
| BC107048.2 | AAI07049.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057847 | Chr.20:42652464 - 44431352 on Build GRCh38 | Loss |
|
| nsv586065 | Chr.20:44261867 - 44505244 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map