Assay Details
Target Gene Details
Entrez Gene ID: | 23339 |
Gene Name: | VPS39, HOPS complex subunit |
Gene Aliases: |
TLP, VAM6, hVam6p |
Location: |
Chr.15:42158701-42208331 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VPS39 | NM_001301138.1 | NP_001288067.1 | ||
| NM_015289.3 | NP_056104.2 | |||
| XM_011521403.2 | XP_011519705.1 | |||
| XM_011521404.2 | XP_011519706.1 | |||
| XM_017022036.1 | XP_016877525.1 | |||
| XM_017022037.1 | XP_016877526.1 | |||
| XM_017022038.1 | XP_016877527.1 | |||
| XM_017022039.1 | XP_016877528.1 | |||
| AB018313.2 | ||||
| AF280814.1 | AAK72222.1 | |||
| AF281052.1 | AAQ05978.1 | |||
| BC015817.2 | AAH15817.2 | |||
| BC068559.1 | AAH68559.1 | |||
| BX537404.1 | CAD97646.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471683 | Chr.15:42042177 - 42210784 on Build GRCh38 | Gain+Loss |
|
| nsv832985 | Chr.15:42079284 - 42272768 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map