Assay Details
Target Gene Details
Entrez Gene ID: | 115703 |
Gene Name: | Rho GTPase activating protein 33 |
Gene Aliases: |
NOMA-GAP, SNX26, TCGAP |
Location: |
Chr.19:35775515-35788822 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP33 | NM_001172630.1 | 21 | 3863 | NP_001166101.1 |
| NM_052948.3 | 21 | 3562 | NP_443180.2 | |
| XM_005258488.1 | 21 | 4041 | XP_005258545.1 | |
| XM_006722999.1 | 21 | 4074 | XP_006723062.1 | |
| XM_006723003.2 | 6 | 2574 | XP_006723066.1 | |
| XM_011526416.1 | 20 | 3929 | XP_011524718.1 | |
| XM_011526417.1 | 20 | 3902 | XP_011524719.1 | |
| XM_011526418.1 | 21 | 3848 | XP_011524720.1 | |
| XM_011526419.1 | 21 | 3815 | XP_011524721.1 | |
| XM_011526420.1 | 20 | 3980 | XP_011524722.1 | |
| AK096338.1 | 21 | 3503 | ||
| AK127255.1 | 21 | 3863 | BAC86902.1 | |
| AL137579.1 | 8 | 2627 | CAB70821.1 | |
| BC014084.2 | 1 | 925 | AAH14084.2 | |
| BC128586.1 | 21 | 2341 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953286 | Chr.19:35747000 - 35868398 on Build GRCh38 | Deletion |
|
| nsv579423 | Chr.19:35730150 - 35792945 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map