Assay Details
Target Gene Details
Entrez Gene ID: | 5017 |
Gene Name: | ovo like transcriptional repressor 1 |
Gene Aliases: |
HOVO1 |
Location: |
Chr.11:65787034-65797219 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OVOL1 | NM_004561.3 | NP_004552.2 | ||
| XM_005274018.3 | XP_005274075.1 | |||
| XM_011545067.2 | XP_011543369.1 | |||
| XM_011545068.2 | XP_011543370.1 | |||
| XM_017017837.1 | XP_016873326.1 | |||
| XM_017017838.1 | XP_016873327.1 | |||
| AF016045.1 | AAB72084.1 | |||
| AK291711.1 | ||||
| BC043146.1 | AAH43146.1 | |||
| BC059408.1 | AAH59408.1 | |||
| BX648725.1 | CAH56127.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759834 | Chr.11:65792422 - 65954892 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map