Assay Details
Target Gene Details
Entrez Gene ID: | 29775 |
Gene Name: | caspase recruitment domain family member 10 |
Gene Aliases: |
BIMP1, CARMA3 |
Location: |
Chr.22:37490362-37519203 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CARD10 | NM_014550.3 | NP_055365.2 | ||
| AF086324.1 | ||||
| AK057334.1 | ||||
| AK074906.1 | BAC11282.1 | |||
| AK296176.1 | ||||
| AY028896.1 | AAK26165.1 | |||
| AY032928.1 | AAK54454.1 | |||
| BC113659.1 | ||||
| BC117403.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829203 | Chr.22:36998126 - 37773322 on Build GRCh38 | Loss |
|
| esv2496764 | Chr.22:37491122 - 37492017 on Build GRCh38 | Insertion |
|
| nsv1057617 | Chr.22:37462633 - 37713698 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map