Assay Details
Target Gene Details
Entrez Gene ID: | 131118 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C19 |
Gene Aliases: |
PAM18, TIM14, TIMM14 |
Location: |
Chr.3:180983709-180989774 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC19 | NM_001190233.1 | 6 | 953 | NP_001177162.1 |
| NM_145261.3 | 6 | 941 | NP_660304.1 | |
| NR_033721.1 | 5 | 889 | ||
| NR_033722.1 | 5 | 861 | ||
| NR_033723.1 | 6 | 886 | ||
| NR_046073.1 | 3 | 735 | ||
| AW207852.1 | ||||
| BC073989.1 | 6 | 886 | AAH73989.1 | |
| BU685935.1 | 1 | 523 | ||
| CB054787.1 |
More Information
Set Membership: |
|

Genomic Map