Assay Details
Target Gene Details
Entrez Gene ID: | 10246 |
Gene Name: | solute carrier family 17 member 2 |
Gene Aliases: |
NPT3 |
Location: |
Chr.6:25912754-25930727 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC17A2 | NM_001286123.1 | 3 | 506 | NP_001273052.1 |
| NM_001286125.1 | 3 | 506 | NP_001273054.1 | |
| NM_005835.3 | 4 | 956 | NP_005826.1 | |
| XM_005248784.2 | 4 | 956 | XP_005248841.1 | |
| XM_006714949.3 | 4 | 693 | XP_006715012.1 | |
| XM_006714950.2 | 3 | 583 | XP_006715013.1 | |
| XM_006714951.1 | 4 | 956 | XP_006715014.1 | |
| XM_017010159.1 | 3 | 846 | XP_016865648.1 | |
| XM_017010160.1 | 4 | 951 | XP_016865649.1 | |
| BC104822.1 | 3 | 479 | AAI04823.1 | |
| BC112033.1 | 3 | 479 | AAI12034.1 | |
| U90544.1 | 4 | 841 | AAB53422.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1023453 | Chr.6:25853602 - 26528022 on Build GRCh38 | Gain |
|
| nsv5229 | Chr.6:25911923 - 25956330 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map