Assay Details
Target Gene Details
Entrez Gene ID: | 283234 |
Gene Name: | coiled-coil domain containing 88B |
Gene Aliases: |
BRLZ, CCDC88, HKRP3, gipie |
Location: |
Chr.11:64340203-64357534 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC88B | NM_032251.5 | 16 | 2857 | NP_115627.6 |
| XM_006718519.3 | 16 | 2869 | XP_006718582.1 | |
| XM_011544957.2 | 16 | 2870 | XP_011543259.1 | |
| XM_011544959.2 | 16 | 2870 | XP_011543261.1 | |
| XM_011544960.2 | 16 | 2869 | XP_011543262.1 | |
| XM_011544962.2 | 3 | 492 | XP_011543264.1 | |
| XM_017017601.1 | 16 | 2871 | XP_016873090.1 | |
| AK074126.1 | 10 | 2291 | BAB84952.1 | |
| AK090436.1 | 16 | 2852 | BAC03417.1 | |
| AK095289.1 | 7 | 1828 | ||
| BC040232.1 | 3 | 414 | ||
| BC140822.1 | 16 | 2846 | ||
| BC140824.1 | 16 | 2846 | ||
| BC141866.1 | 16 | 2836 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv555193 | Chr.11:64339671 - 64353102 on Build GRCh38 | Loss |
|
| nsv951010 | Chr.11:63942829 - 64352128 on Build GRCh38 | Deletion |
|
| nsv358 | Chr.11:64325464 - 64359765 on Build GRCh38 | Insertion |
|
| nsv832186 | Chr.11:64204788 - 64402587 on Build GRCh38 | Loss |
|
| dgv13n64 | Chr.11:64329761 - 64362250 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map