Assay Details
Target Gene Details
Entrez Gene ID: | 2355 |
Gene Name: | FOS like 2, AP-1 transcription factor subunit |
Gene Aliases: |
FRA2 |
Location: |
Chr.2:28392759-28417312 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOSL2 | NM_005253.3 | 4 | 2493 | NP_005244.1 |
| XM_006711976.2 | 4 | 2697 | XP_006712039.1 | |
| XM_006711977.3 | 4 | 3550 | XP_006712040.1 | |
| XM_017003737.1 | 4 | 3206 | XP_016859226.1 | |
| AK026959.1 | 1 | 228 | BAB15603.1 | |
| BX647822.1 | 4 | 1669 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1006830 | Chr.2:28331908 - 28419880 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map