Assay Details
Target Gene Details
Entrez Gene ID: | 1047 |
Gene Name: | calmegin |
Gene Aliases: |
- |
Location: |
Chr.4:140388453-140427661 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLGN | NM_001130675.1 | 13 | 1639 | NP_001124147.1 |
| NM_004362.2 | 12 | 1555 | NP_004353.1 | |
| AK093096.1 | 12 | 2118 | ||
| AK290819.1 | 12 | 1552 | ||
| AK299243.1 | 11 | 1344 | ||
| AK302149.1 | 8 | 927 | ||
| BC028357.2 | 12 | 1509 | AAH28357.1 | |
| D86322.1 | 12 | 1522 | BAA22590.1 |
More Information
Set Membership: |
|

Genomic Map