Assay Details
Target Gene Details
Entrez Gene ID: | 84720 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class O |
Gene Aliases: |
HPMRS2 |
Location: |
Chr.9:35085493-35096601 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIGO | NM_001201484.1 | 11 | 2181 | NP_001188413.1 |
| NM_032634.3 | 9 | 3334 | NP_116023.2 | |
| NM_152850.3 | 10 | 1729 | NP_690577.2 | |
| XM_005251619.3 | 9 | 2959 | XP_005251676.1 | |
| XM_017015222.1 | 10 | 3451 | XP_016870711.1 | |
| XM_017015223.1 | 10 | 2110 | XP_016870712.1 | |
| XM_017015224.1 | 10 | 2374 | XP_016870713.1 | |
| AK074064.1 | 5 | 3943 | BAB84890.1 | |
| AK090433.1 | 4 | 5148 | BAC03414.1 | |
| AL833956.1 | 9 | 3367 | CAD38806.1 | |
| AY358472.1 | 8 | 2940 | AAQ88836.1 | |
| BC001030.2 | 7 | 1059 | AAH01030.1 | |
| BC013987.2 | 8 | 1145 | AAH13987.1 | |
| BC029271.1 | 8 | 2605 | AAH29271.1 | |
| BC036916.1 | 9 | 1694 | AAH36916.1 | |
| BC065282.1 | 11 | 1852 | AAH65282.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1030303 | Chr.9:35084357 - 35506646 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map