Assay Details
Target Gene Details
Entrez Gene ID: | 114960 |
Gene Name: | testis specific 13 |
Gene Aliases: |
- |
Location: |
Chr.7:130668646-130687432 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSGA13 | NM_001304968.1 | NP_001291897.1 | ||
| NM_052933.3 | NP_443165.1 | |||
| AF294347.1 | AAK97089.1 | |||
| AF431815.1 | AAP97305.1 | |||
| AK093329.1 | ||||
| BC038415.1 | AAH38415.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831132 | Chr.7:130511850 - 130727326 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map