Assay Details
Target Gene Details
Entrez Gene ID: | 84276 |
Gene Name: | nicolin 1 |
Gene Aliases: |
- |
Location: |
Chr.3:49422333-49429324 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NICN1 | NM_032316.3 | NP_115692.1 | ||
| AF538150.1 | AAN52734.1 | |||
| AJ299740.1 | CAC82516.1 | |||
| AK094248.1 | ||||
| AK301846.1 | ||||
| AK313073.1 | ||||
| BC005050.1 | AAH05050.1 | |||
| BC050653.1 | AAH50653.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834687 | Chr.3:49411199 - 49594355 on Build GRCh38 | Loss |
|

Genomic Map