Assay Details
Target Gene Details
Entrez Gene ID: | 60494 |
Gene Name: | coiled-coil domain containing 81 |
Gene Aliases: |
- |
Location: |
Chr.11:86374736-86423109 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC81 | NM_001156474.1 | 1 | 57 | NP_001149946.1 |
| NM_021827.4 | 1 | 57 | NP_068599.3 | |
| XM_005274161.1 | 1 | 57 | XP_005274218.1 | |
| AK131331.1 | 1 | 60 | BAD18491.1 |

Genomic Map