Assay Details
Target Gene Details
Entrez Gene ID: | 9796 |
Gene Name: | phytanoyl-CoA 2-hydroxylase interacting protein |
Gene Aliases: |
DYRK1AP3, PAHX-AP, PAHXAP1 |
Location: |
Chr.8:22219703-22232338 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHYHIP | NM_001099335.1 | 6 | 2673 | NP_001092805.1 |
| NM_014759.3 | 5 | 2551 | NP_055574.3 | |
| XM_006716416.1 | XP_006716479.1 | |||
| XM_017014102.1 | 3 | 2816 | XP_016869591.1 | |
| AK098743.1 | ||||
| BC034034.1 | 5 | 2304 | AAH34034.1 | |
| BC035940.1 | 5 | 2304 | AAH35940.1 | |
| D87463.1 | 6 | 2382 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470205 | Chr.8:22191277 - 22256052 on Build GRCh38 | Loss |
|
| dgv12054n54 | Chr.8:22045805 - 22251098 on Build GRCh38 | Loss |
|
| nsv831265 | Chr.8:22199688 - 22366918 on Build GRCh38 | Loss |
|
| nsv831264 | Chr.8:22093700 - 22328040 on Build GRCh38 | Gain+Loss |
|
| esv2759604 | Chr.8:20675533 - 23254096 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73551785] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map