Assay Details
Target Gene Details
Entrez Gene ID: | 11016 |
Gene Name: | activating transcription factor 7 |
Gene Aliases: |
ATFA |
Location: |
Chr.12:53507856-53626415 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATF7 | NM_001130060.1 | NP_001123532.1 | ||
| NM_001206682.1 | NP_001193611.1 | |||
| NM_001206683.1 | NP_001193612.1 | |||
| NM_006856.2 | NP_006847.1 | |||
| NR_073163.1 | ||||
| XM_005268587.3 | XP_005268644.1 | |||
| XM_017018722.1 | XP_016874211.1 | |||
| AA679547.1 | ||||
| AK298853.1 | ||||
| AK302907.1 | ||||
| AK313507.1 | ||||
| BC042363.1 | AAH42363.1 | |||
| BC136302.1 | ||||
| BC140006.1 | ||||
| DB047160.1 | ||||
| DB490927.2 | ||||
| X52943.1 | CAA37118.1 | |||
| X57197.1 | CAA40483.1 |
More Information
Set Membership: |
|

Genomic Map