Assay Details
Target Gene Details
Entrez Gene ID: | 5083 |
Gene Name: | paired box 9 |
Gene Aliases: |
STHAG3 |
Location: |
Chr.14:36657568-36677807 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAX9 | NM_006194.3 | NP_006185.1 | ||
| AB248958.1 | BAE79273.1 | |||
| AK313627.1 | ||||
| BC001159.1 | AAH01159.1 | |||
| U59628.1 | AAD09487.1 | |||
| X92850.1 | CAA63436.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952826 | Chr.14:36653796 - 36663795 on Build GRCh38 | Deletion |
|
| nsv819585 | Chr.14:36661422 - 36669449 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map