Assay Details
Target Gene Details
Entrez Gene ID: | 8331 |
Gene Name: | histone cluster 1, H2aj |
Gene Aliases: |
H2A/E, H2AFE, dJ160A22.4 |
Location: |
Chr.6:27814302-27814740 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HIST1H2AJ | NM_021066.2 | 1 | 373 | NP_066544.1 |
| BC066232.1 | AAH66232.1 | |||
| BC066233.1 | AAH66233.1 | |||
| BC066234.1 | AAH66234.1 | |||
| BC066235.1 | AAH66235.1 | |||
| BC066236.1 | AAH66236.1 | |||
| BC066237.1 | AAH66237.1 | |||
| BC133048.1 | 1 | 462 | ||
| BC133050.1 | 1 | 462 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3890803 | Chr.6:27771601 - 27814760 on Build GRCh38 | Loss |
|
| nsv1017057 | Chr.6:27777998 - 27821905 on Build GRCh38 | Gain |
|
| nsv830618 | Chr.6:27625495 - 27821827 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map