Assay Details
Target Gene Details
Entrez Gene ID: | 386724 |
Gene Name: | adhesion molecule with Ig like domain 3 |
Gene Aliases: |
ALI3, AMIGO-3 |
Location: |
Chr.3:49716834-49719805 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AMIGO3 | NM_198722.2 | 1 | 1895 | NP_942015.1 |
| AY237003.1 | AAO48944.1 | |||
| AY358134.1 | 1 | 1780 | AAQ88501.1 | |
| BC101363.1 | 1 | 1716 | AAI01364.1 | |
| BC101364.1 | 1 | 1716 | AAI01365.1 | |
| BC101365.2 | 1 | 1716 | AAI01366.1 | |
| BC110418.1 | 1 | 1785 |
Target Gene Details
Entrez Gene ID: | 63891 |
Gene Name: | ring finger protein 123 |
Gene Aliases: |
FP1477, KPC1 |
Location: |
Chr.3:49688753-49721529 on Build GRCh38 |
Assay Gene Location: | Within Intron 37 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF123 | NM_022064.4 | NP_071347.2 | ||
| NR_135218.1 | ||||
| XM_006713285.1 | XP_006713348.1 | |||
| XM_011533995.1 | XP_011532297.1 | |||
| XM_017007018.1 | XP_016862507.1 | |||
| AF370367.1 | AAQ15203.1 | |||
| AK022627.1 | BAB14139.1 | |||
| AK026968.1 | BAB15607.1 | |||
| AK090632.1 | ||||
| AK096353.1 | ||||
| AK126883.1 | ||||
| AL136729.1 | CAB66663.2 | |||
| AL832034.1 | ||||
| AL832138.1 | ||||
| AL832344.1 | ||||
| AY744152.1 | AAU93470.1 | |||
| BC041145.1 | AAH41145.1 | |||
| BC057392.1 | AAH57392.2 | |||
| BC088801.1 | AAH88801.1 | |||
| BC130632.1 | ||||
| CR627051.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508217 | Chr.3:49578013 - 49732463 on Build GRCh38 | Deletion |
|
| nsv482082 | Chr.3:49717532 - 49719690 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map