Assay Details
Target Gene Details
Entrez Gene ID: | 3431 |
Gene Name: | SP110 nuclear body protein |
Gene Aliases: |
IFI41, IFI75, IPR1, VODI |
Location: |
Chr.2:230165466-230225729 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SP110 | NM_004509.3 | NP_004500.3 | ||
| NM_080424.2 | NP_536349.2 | |||
| XM_005246525.3 | XP_005246582.1 | |||
| XM_006712487.3 | XP_006712550.1 | |||
| XM_006712489.3 | XP_006712552.1 | |||
| XM_011511088.2 | XP_011509390.1 | |||
| XM_011511089.2 | XP_011509391.1 | |||
| XM_011511090.2 | XP_011509392.1 | |||
| XM_011511091.2 | XP_011509393.1 | |||
| XM_011511092.2 | XP_011509394.1 | |||
| XM_017003968.1 | XP_016859457.1 | |||
| AF280095.1 | AAG09826.1 | |||
| AK026488.1 | ||||
| AK128274.1 | 1 | 524 | ||
| L22343.1 | AAD13402.1 |
More Information
Set Membership: |
|

Genomic Map