Assay Details
Target Gene Details
Entrez Gene ID: | 4892 |
Gene Name: | nebulin related anchoring protein |
Gene Aliases: |
N-RAP |
Location: |
Chr.10:113588824-113664070 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 38 - Exon 38 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NRAP | NM_001261463.1 | NP_001248392.1 | ||
| NM_001322945.1 | NP_001309874.1 | |||
| NM_006175.4 | NP_006166.3 | |||
| NM_198060.3 | NP_932326.2 | |||
| XM_005269864.2 | XP_005269921.1 | |||
| XM_005269865.2 | XP_005269922.1 | |||
| XM_005269867.2 | XP_005269924.1 | |||
| XM_006717870.2 | XP_006717933.1 | |||
| XM_011539832.2 | XP_011538134.1 | |||
| AF542553.1 | AAQ09536.1 | |||
| AK056605.1 | BAB71231.1 | |||
| AK096875.1 | ||||
| AK096886.1 | BAC04884.1 | |||
| AL831980.2 | CAD89899.1 | |||
| AL832025.2 | CAD89910.1 | |||
| AL832330.2 | CAD38623.1 | |||
| AL832457.3 | CAD89998.1 | |||
| AY081943.2 | AAL99185.2 | |||
| AY177620.1 | AAO47073.1 | |||
| AY177621.1 | AAO47074.1 | |||
| BC126407.1 | ||||
| BC143751.1 | ||||
| BX640682.2 | CAE45811.1 | |||
| BX640730.1 | CAE45846.1 | |||
| BX641052.1 | CAE46027.1 | |||
| BX647658.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3624603 | Chr.10:113581857 - 113745416 on Build GRCh38 | Gain |
|
| nsv1048819 | Chr.10:113499347 - 113757619 on Build GRCh38 | Gain |
|
| nsv7555 | Chr.10:113585355 - 113630190 on Build GRCh38 | Deletion |
|
| esv3624604 | Chr.10:113582786 - 113669854 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map