Assay Details
Target Gene Details
Entrez Gene ID: | 7108 |
Gene Name: | transmembrane 7 superfamily member 2 |
Gene Aliases: |
ANG1, DHCR14A, NET47 |
Location: |
Chr.11:65111854-65116235 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TM7SF2 | NM_001277233.1 | NP_001264162.1 | ||
| NM_003273.3 | NP_003264.2 | |||
| NR_102367.1 | ||||
| AF023676.1 | AAC21450.1 | |||
| AF096304.1 | AAD09765.1 | |||
| AK290935.1 | ||||
| AK296441.1 | ||||
| BC009052.1 | AAH09052.1 | |||
| BC012857.1 | AAH12857.1 | |||
| BC038353.1 | AAH38353.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526181 | Chr.11:64870569 - 65489858 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map