Assay Details
Target Gene Details
Entrez Gene ID: | 9519 |
Gene Name: | TATA-box binding protein like 1 |
Gene Aliases: |
MGC:8389, MGC:9620, STUD, TLF, TLP, TRF2 |
Location: |
Chr.6:133952170-133987500 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBPL1 | NM_001253676.1 | NP_001240605.1 | ||
| NM_004865.3 | 1 | 87 | NP_004856.1 | |
| XM_017011513.1 | XP_016867002.1 | |||
| AB020881.1 | BAA75218.1 | |||
| AF130312.1 | AAD24800.1 | |||
| AF136570.1 | AAD28785.1 | |||
| AK292320.1 | ||||
| AK313645.1 | ||||
| BC000381.2 | AAH00381.1 | |||
| BC017559.2 | AAH17559.1 | |||
| BI560657.1 | 1 | 92 | ||
| BI828497.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1832e212 | Chr.6:133945117 - 133957026 on Build GRCh38 | Loss |
|
| esv34122 | Chr.6:133767881 - 134057114 on Build GRCh38 | Loss |
|
| esv3571405 | Chr.6:133939645 - 133965091 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs113696984] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map