Assay Details
Target Gene Details
Entrez Gene ID: | 146206 |
Gene Name: | capping protein regulator and myosin 1 linker 2 |
Gene Aliases: |
CARMIL2b, LRRC16C, RLTPR |
Location: |
Chr.16:67645004-67657569 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CARMIL2 | NM_001013838.1 | 21 | 2086 | NP_001013860.1 |
| NM_001317026.1 | 22 | 1978 | NP_001303955.1 | |
| XM_011522874.1 | 22 | 2116 | XP_011521176.1 | |
| XM_011522875.2 | 22 | 2116 | XP_011521177.2 | |
| XM_017022953.1 | 22 | 2116 | XP_016878442.1 | |
| AB113647.1 | 21 | 2086 | BAD26751.1 | |
| AK090421.1 | 3 | 1753 | BAC03402.1 | |
| AL832446.1 | 20 | 1917 | ||
| FJ026014.1 | 22 | 1858 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522852 | Chr.16:67153308 - 68071571 on Build GRCh38 | Gain |
|
| nsv952046 | Chr.16:67628798 - 67678697 on Build GRCh38 | Deletion |
|
| nsv524363 | Chr.16:67136529 - 68071571 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map