Assay Details
Target Gene Details
Entrez Gene ID: | 85440 |
Gene Name: | dedicator of cytokinesis 7 |
Gene Aliases: |
EIEE23, ZIR2 |
Location: |
Chr.1:62454726-62688368 on Build GRCh38 |
Assay Gene Location: | Within Exon 53 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DOCK7 | NM_001271999.1 | 49 | 6529 | NP_001258928.1 |
| NM_001272000.1 | 49 | 6442 | NP_001258929.1 | |
| NM_001272001.1 | 48 | 6436 | NP_001258930.1 | |
| NM_033407.3 | 49 | 6469 | NP_212132.2 | |
| XM_005271292.2 | 50 | 6525 | XP_005271349.1 | |
| XM_011542326.2 | 50 | 6552 | XP_011540628.1 | |
| XM_011542327.2 | 49 | 6546 | XP_011540629.1 | |
| XM_011542328.2 | 49 | 6537 | XP_011540630.1 | |
| XM_017002639.1 | 48 | 6453 | XP_016858128.1 | |
| AB051558.1 | 30 | 3945 | BAB21862.1 | |
| AK055296.1 | 2 | 1738 | ||
| AK055493.1 | 16 | 2086 | BAB70933.1 | |
| AK055905.1 | 21 | 2961 | BAB71042.1 | |
| AK091479.1 | 23 | 2936 | ||
| AK125049.1 | 35 | 4614 | BAC86032.1 | |
| AK299737.1 | ||||
| BC016392.1 | 18 | 2433 | ||
| BC112387.1 | 9 | 1188 | ||
| BG190976.1 | 1 | 82 | ||
| BG205481.1 | 1 | 83 | ||
| BG207092.1 | ||||
| DQ118679.1 | AAZ38451.1 | |||
| DQ118680.1 | AAZ38452.1 | |||
| DQ341187.1 | 49 | 6436 |
More Information
Set Membership: |
|

Genomic Map