Assay Details
Target Gene Details
Entrez Gene ID: | 134492 |
Gene Name: | NudC domain containing 2 |
Gene Aliases: |
NudCL2 |
Location: |
Chr.5:163453580-163460137 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUDCD2 | NM_145266.4 | 1 | 224 | NP_660309.1 |
| AK311957.1 | 1 | 187 | ||
| BC017934.1 | 1 | 224 | AAH17934.1 | |
| BX538290.1 | 1 | 225 | CAD98085.1 | |
| FJ215684.1 | 1 | 164 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1018494 | Chr.5:163390438 - 163476503 on Build GRCh38 | Gain |
|
| nsv1017056 | Chr.5:163328870 - 163804011 on Build GRCh38 | Gain |
|
| nsv830538 | Chr.5:163377440 - 163541647 on Build GRCh38 | Gain+Loss |
|
| nsv1029437 | Chr.5:163104166 - 164109116 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map