Assay Details
Target Gene Details
Entrez Gene ID: | 4759 |
Gene Name: | neuraminidase 2 |
Gene Aliases: |
SIAL2 |
Location: |
Chr.2:233032672-233035057 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEU2 | NM_005383.2 | 2 | 929 | NP_005374.2 |
| BC069151.1 | 2 | 929 | AAH69151.1 | |
| BC107053.2 | 2 | 929 | AAI07054.1 |
More Information
Set Membership: |
|

Genomic Map