Assay Details
Target Gene Details
Entrez Gene ID: | 5498 |
Gene Name: | protoporphyrinogen oxidase |
Gene Aliases: |
PPO, V290M, VP |
Location: |
Chr.1:161165728-161178277 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 13 - Intron 13 |
More Information
Set Membership: |
|

Genomic Map